missing translation for 'onlineSavingsMsg'
Learn More

GAT3 Antibody, R&D Systems™

Product Code. 18603822 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18603822 100 μg 100µL
18684402 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18603822 Supplier Novus Biologicals Supplier No. NBP297671100ul
  • ONLINE EXCLUSIVE PRICE
    345.81 €
    307.65 € / 100µL
    Save 38.16 €
    11% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

GAT3 Polyclonal specifically detects GAT3 in Human samples. It is validated for Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAT3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.0)
Gene Alias GABA Transporter 3, GABT3, GAT-3, GAT3sodium- and chloride-dependent GABA transporter 3, GAT4, solute carrier family 6 (neurotransmitter transporter, GABA), member 11, Solute carrier family 6 member 11
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human GAT3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6538
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.