missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, Mikromol™

Product Code. 17969454
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17969454 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17969454 Supplier Mikromol Supplier No. MM0169.000250
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Glutamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Glutamine
CAS 56-85-9
Analyte or Component Names L-Glutamine
Shipping Condition Room Temperature
Molecular Formula C5 H10 N2 O3
Certifications/Compliance ISO 17034
Synonym L-Glutamine, (2S)-2,5-Diamino-5-oxopentanoic Acid, Aspartic Acid Imp. F (Pharmeuropa), 2-Aminoglutaramic acid, Acustasin, Aesgen 14, Cebrogen, Glumin, Glumin (amino acid), Glutamic acid 5-amide, Glutamic acid amide, Glutamine, L-(+)-Glutamine, L-2-Aminoglutaramidic acid, Levoglutamide, NSC 27421, (S)-2,5-Diamino-5-oxopentanoic acid, Saforis, Stimulina, L-Glutamic acid gamma-amide, gamma-Glutamine, Asparagine Monohydrate Impurity E, Aspartic Acid Impurity F, Magnesium Aspartate Dihydrate Impurity G, Asparagine Impurity E, Magnesium Aspartate Impurity G
InChI Formula InChI=1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1
SMILES N[C@@H](CCC(=O)N)C(=O)O
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
Molecular Weight (g/mol) 146.14
Quantity 250 mg
Formula Weight 146.0691 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.