missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Methyl Ester, Mikromol™

Product Code. 30457519
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457519 4 x 25 mg -
30050020 100 mg 100mg
30050762 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457519 Supplier Mikromol Supplier No. MM0634.0400254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Tyrosine Methyl Ester, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Tyrosine Methyl Ester
CAS 1080-06-4
Analyte or Component Names L-Tyrosine Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C10 H13 N O3
Certifications/Compliance ISO 17025
Synonym L-Tyrosine methyl ester, (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid methyl ester, Methyl (2S)-2-amino-3-(4-hydroxyphenyl)propionate, Methyl (2S)-3-(4-hydroxyphenyl)-2-aminopropionate, Methyl (S)-2-amino-3-(4-hydroxyphenyl)propanoate, Methyl L-tyrosinate, Methyl tyrosinate, S-Tyrosine methyl ester, Tyrosine methyl ester
InChI Formula InChI=1S/C10H13NO3/c1-14-10(13)9(11)6-7-2-4-8(12)5-3-7/h2-5,9,12H,6,11H2,1H3/t9-/m0/s1
Solution Type Neat
SMILES COC(=O)[C@@H](N)Cc1ccc(O)cc1
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol) 195.22
Quantity 4 x 25 mg
Formula Weight 195.0895 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.