missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (CL12988), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18553-100ul
This item is not returnable.
View return policy
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| L1CAM | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12988 | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| This antibody was developed against a sythetic peptide for P32004. Actual immunogen sequence is proprietary. | |
| 100 μg | |
| Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
| 3897 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction