missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L1CAM Antibody (CL12988), Novus Biologicals™
Mouse Monoclonal Antibody
429.00 € - 611.00 €
Specifications
| Antigen | L1CAM |
|---|---|
| Clone | CL12988 |
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683254
|
Novus Biologicals
NBP3-18553-100ul |
100 μg |
611.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688654
|
Novus Biologicals
NBP3-18553-25ul |
25 μg |
429.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L1CAM Monoclonal antibody specifically detects L1CAM in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| L1CAM | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1 | |
| This antibody was developed against a sythetic peptide for P32004. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12988 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Glia Markers, Neuroscience, Phospho Specific | |
| PBS, pH 7.2, 40% glycerol | |
| 3897 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title