missing translation for 'onlineSavingsMsg'
Learn More
Learn More
La Crosse Virus Nucleocapsid Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41225-0.025mg
This item is not returnable.
View return policy
Description
La Crosse Virus Nucleocapsid Polyclonal specifically detects La Crosse Virus Nucleocapsid in Virus samples. It is validated for ELISA.
Specifications
| La Crosse Virus Nucleocapsid | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| 26 kDa | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| ELISA | |
| 1 mg/mL | |
| ELISA 1 μg/mL | |
| LACVsSgp1, N, Nucleocapsid protein, Nucleoprotein, Protein N | |
| Antibody was raised against an 18 amino acid peptide near the center of the La Crosse virus nucleocapsid. The immunogen is located within amino acids 90 - 140 of La Crosse Virus Nucleocapsid. Amino Acid Squence: LDQYNENDDESQHELIRT | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction