missing translation for 'onlineSavingsMsg'
Learn More
Learn More
La Crosse Virus Nucleocapsid Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00 € - 544.00 €
Specifications
| Antigen | La Crosse Virus Nucleocapsid |
|---|---|
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677636
|
Novus Biologicals
NBP2-41225-0.025mg |
0.025 mg |
189.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18156318
|
Novus Biologicals
NBP2-41225 |
0.1 mg |
544.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
La Crosse Virus Nucleocapsid Polyclonal specifically detects La Crosse Virus Nucleocapsid in Virus samples. It is validated for ELISA.Specifications
| La Crosse Virus Nucleocapsid | |
| ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Antibody was raised against an 18 amino acid peptide near the center of the La Crosse virus nucleocapsid. The immunogen is located within amino acids 90 - 140 of La Crosse Virus Nucleocapsid. Amino Acid Squence: LDQYNENDDESQHELIRT | |
| Primary | |
| 26 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| LACVsSgp1, N, Nucleocapsid protein, Nucleoprotein, Protein N | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title