missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Negative Control for Mouse IgG2b Ab-1 with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Mouse IgG2b Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ NC-1391-PABX

 View more versions of this product

Product Code. 12682677

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This mouse monoclonal IgG2b was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG2b does not react with any antigen of human or animal cells. The monoclonal IgG2b was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG2b should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG2b antibodies.

Host Species: Mouse

Clone: NCG2B.01

Isotype: IgG2b/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Negative Control for Mouse IgG2b Ab-1
Monoclonal
1mg/mL
Mouse
RUO
Primary
Concentrated
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
NCG2B.01
Unconjugated
200 μL
Negative Controls
All species
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.