missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Negative Control for Mouse IgG2b Ab-1 with BSA and azide

Take advantage of a quality product that delivers accurate results with the Epredia™ Negative Control for Mouse IgG2b Ab-1, Mouse Monoclonal Antibody.

132.60 € - 443.70 €

Specifications

Antigen Negative Control for Mouse IgG2b Ab-1
Clone NCG2B.01
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12652677
View Documents
Epredia™
NC-1391-P0
100 μL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12672677
View Documents
Epredia™
NC-1391-P1ABX
100 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12682677
View Documents
Epredia™
NC-1391-PABX
200 μL 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12662677
View Documents
Epredia™
NC-1391-P1
500 μL 200μg/mL
ONLINE EXCLUSIVE PRICE
369.00 €
308.77 € / 500µL
Save 60.23 €
16% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12642677
View Documents
Epredia™
NC-1391-P
1 mL 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12602687
View Documents
Epredia™
NC-1391-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This mouse monoclonal IgG2b was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG2b does not react with any antigen of human or animal cells. The monoclonal IgG2b was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG2b should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG2b antibodies.

Host Species: Mouse

Clone: NCG2B.01

Isotype: IgG2b/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Negative Control for Mouse IgG2b Ab-1
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
All species
Primary
NCG2B.01
Monoclonal
Mouse
Negative Controls
IgG2b κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.