missing translation for 'onlineSavingsMsg'
Learn More

Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-394-B0

 View more versions of this product

Product Code. 12601307

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

NGF is important for the development, differentiation, and survival of variety of neuronal and non-neuronal cells. Its action is mediated by binding two distinct receptors, the high affinity p140 and low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR).

Host Species: Mouse

Clone: NGFR5

Isotype: IgG1/κ

Species Reactivity: Human, Monkey (Cebus apella, Macaca fascicularis, mulatta, and nemestrina), Baboon (Papio anubis and Cynocephalus), Cat, Rabbit, Ferret. Does not react with mouse and rat.

Epitope: aa 1-160

Immunogen: NGFR from A875 melanoma cells

Molecular Weight: 75kDa

Positive Control: Melanoma

Cellular Localization: Cell surface and cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits Binding of NGF to Melanoma Cells

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Neurotrophin Receptor/NGF-Receptor (p75superscriptNGFRsuperscript) Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Neuroscience
Feline, Ferret, Human, Non-human Primate, Rabbit
IgG2a κ
Flow Cytometry
NGFR5
Biotin
NGFR from A875 melanoma cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.