missing translation for 'onlineSavingsMsg'
Learn More

Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1 Mouse Monoclonal Antibody, Epredia™

Epredia™ Neurotrophin Receptor/NGF-Receptor (p75NGFR) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

149.94 € - 683.40 €

Specifications

Antigen Neurotrophin Receptor/NGF-Receptor (p75superscriptNGFRsuperscript) Ab-1
Clone NGFR5
Applications Flow Cytometry
Classification Monoclonal
Form Concentrated
View More Specs

Products 7
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
12601307
View Documents
Epredia
MS-394-B0
100 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12611307
View Documents
Epredia
MS-394-B1
500 μL Biotin 200μg/mL
ONLINE EXCLUSIVE PRICE
650.00 €
535.03 € / 500µL
Save 114.97 €
17% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12631307
View Documents
Epredia
MS-394-P0
100 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12641307
View Documents
Epredia
MS-394-P1
500 μL Unconjugated 200μg/mL
ONLINE EXCLUSIVE PRICE
487.00 €
410.23 € / 500µL
Save 76.77 €
15% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12621307
View Documents
Epredia
MS-394-P
1 mL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12651307
View Documents
Epredia
MS-394-P1ABX
100 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12681307
View Documents
Epredia
MS-394-PABX
200 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

NGF is important for the development, differentiation, and survival of variety of neuronal and non-neuronal cells. Its action is mediated by binding two distinct receptors, the high affinity p140 and low affinity Nerve Growth Factor (NGF) Receptor (p75NGFR) or Neurotrophin Receptor (p75NTR).

Host Species: Mouse

Clone: NGFR5

Isotype: IgG1/κ

Species Reactivity: Human, Monkey (Cebus apella, Macaca fascicularis, mulatta, and nemestrina), Baboon (Papio anubis and Cynocephalus), Cat, Rabbit, Ferret. Does not react with mouse and rat.

Epitope: aa 1-160

Immunogen: NGFR from A875 melanoma cells

Molecular Weight: 75kDa

Positive Control: Melanoma

Cellular Localization: Cell surface and cytoplasmic

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Not suitable)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits Binding of NGF to Melanoma Cells

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Neurotrophin Receptor/NGF-Receptor (p75superscriptNGFRsuperscript) Ab-1
Flow Cytometry
Concentrated
RUO
Feline, Ferret, Human, Non-human Primate, Rabbit
IgG2a κ
NGFR5
Monoclonal
Mouse
Neuroscience
NGFR from A875 melanoma cells
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.