missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labetalol Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Labetalol Hydrochloride |
| CAS | 32780-64-6 |
| Analyte or Component Names | Labetalol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 N2 O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, hydrochloride (1:1), Benzamide, 2-hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride (9CI), Salicylamide, 5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]-, monohydrochloride (8CI), AH 5158 hydrochloride, AH 5158A, Amipress, Ipolab, Labelol, Labetalol hydrochloride, Labracol, NSC 290312, Normodyne, Presdate, Pressalolo, Sch 15719W, Trandate, Vescal |
| InChI Formula | InChI=1S/C19H24N2O3.ClH/c1-13(7-8-14-5-3-2-4-6-14)21-12-18(23)15-9-10-17(22)16(11-15)19(20)24;/h2-6,9-11,13,18,21-23H,7-8,12H2,1H3,(H2,20,24);1H |
| Solution Type | Neat |
| SMILES | Cl.CC(CCc1ccccc1)NCC(O)c2ccc(O)c(c2)C(=O)N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction