missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lacosamide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Lacosamide
Specifications
Specifications
| Chemical Name or Material | Lacosamide |
| CAS | 175481-36-4 |
| Analyte or Component Names | Lacosamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 N2 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2R)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)-propanamide, Propanamide, 2-(acetylamino)-3-methoxy-N-(phenylmethyl)-, (R)-, (2R)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)propanamide, (2R)-2-Acetamido-N-benzyl-3-methoxypropanamide, (R)-2-Acetamido-N-benzyl-3-methoxypropionamide, ADD 243037, Erlosamide, Harkoseride, Lacosamide, Lacoset, Lacoset 50, SPM 927, Vimpat |
| InChI Formula | InChI=1S/C13H18N2O3/c1-10(16)15-12(9-18-2)13(17)14-8-11-6-4-3-5-7-11/h3-7,12H,8-9H2,1-2H3,(H,14,17)(H,15,16)/t12-/m1/s1 |
| SMILES | COC[C@@H](NC(=O)C)C(=O)NCc1ccccc1 |
| IUPAC Name | (2R)-2-acetamido-N-benzyl-3-methoxypropanamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction