missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactase Antibody (4E10 10.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50513-0.025ml
This item is not returnable.
View return policy
Description
Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Lactase | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
| Affinity purified human lactase | |
| 0.025 mL | |
| Cancer, Cellular Signaling, metabolism | |
| 3938 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 4E10 10.2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction