missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactase Antibody (4E10 10.2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
201.00 € - 351.00 €
Specifications
| Antigen | Lactase |
|---|---|
| Clone | 4E10 10.2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616517
|
Novus Biologicals
NBP2-50513-0.025ml |
0.025 mL |
201.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689696
|
Novus Biologicals
NBP2-50513 |
0.1 mL |
351.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Lactase | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Signaling, metabolism | |
| PBS | |
| 3938 | |
| IgG1 | |
| Protein A purified |
| 4E10 10.2 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
| Affinity purified human lactase | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title