missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin alpha 3 Antibody (CL3112) [mFluor Violet 610 SE], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Laminin alpha 3 Monoclonal antibody specifically detects Laminin alpha 3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Laminin alpha 3 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL3112 |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BM600, BM600 150kD subunit, BM600-150kDa, E170, epiligrin, Epiligrin 170 kDa subunit, epiligrin alpha 3 subunit, Epiligrin subunit alpha, kalinin 165kD subunit, Kalinin subunit alpha, kalinin-165kDa, lama3a, laminin subunit alpha-3, laminin, alpha 3, laminin, alpha 3 (nicein (150kD), kalinin (165kD), BM600 (150kD), epilegrin), laminin-5 alpha 3 chain, Laminin-5 subunit alpha, Laminin-6 subunit alpha, Laminin-7 subunit alpha, LAMNA, LOCS, nicein 150kD subunit, Nicein subunit alpha, nicein-150kDa |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from Q16787, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?