missing translation for 'onlineSavingsMsg'
Learn More

Lamivudeine 100 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer

Product Code. 30328071
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328071 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328071 Supplier Dr. Ehrenstorfer Supplier No. DREA14591800WL100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lamivudine
CAS 134678-17-4
Analyte or Component Names Lamivudine
Shipping Condition Room Temperature
Molecular Formula C8 H11 N3 O3 S
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Lamivudine, 2(1H)-Pyrimidinone, 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, 2(1H)-Pyrimidinone, 4-amino-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, (2R-cis)-, 1,3-Oxathiolane, 2(1H)-pyrimidinone deriv., (-)-2'-Deoxy-3'-thiacytidine, (-)-BCH 189, 2-Hydroxymethyl-5-(cytosin-1-yl)-1,3-oxathiolane, 3TC, BCH 189, (-)-, Epivir, Epivir HBV, GR 109714X, Hepitec, Heptodin, Heptovir, L-SddC, Lamivir, Virolam, Zeffix, Zefix, cis(-)-2-Hydroxymethyl-5-(cytosin-1-yl)-1,3-oxathiolane, beta-L-2',3'-Dideoxy-3'-thiacytidine, beta-L-3'-Thia-2',3'-dideoxycytidine, GR 109714X, 3TC, GR 111720X, GR 135522X, Lamivudine GR109714X, BP Zidovudine and Lamivudine Impurity Standard, Ph Eur Lamivudine for system suitability 1, BP Lamivudine Impurity Standard, Ph Eur Lamivudine for system suitability 2
InChI Formula InChI=1S/C8H11N3O3S/c9-5-1-2-11(8(13)10-5)6-4-15-7(3-12)14-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7+/m0/s1
SMILES NC1=NC(=O)N(C=C1)[C@@H]2CS[C@H](CO)O2
IUPAC Name 4-amino-1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
Solvent or Matrix Acetonitrile/Water
Molecular Weight (g/mol) 229.26
Quantity 1 mL
Formula Weight 229.0521 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.