missing translation for 'onlineSavingsMsg'
Learn More

Lamotrigine EP Impurity A (Lamotrigine USP Related Compound C), Mikromol™

Product Code. 18033780
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18033780 100 mg 100mg
18000170 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18033780 Supplier Mikromol Supplier No. MM0922.01
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Lamotrigine EP Impurity A (Lamotrigine USP Related Compound C)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one
CAS 252186-78-0
Analyte or Component Names 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one
Shipping Condition Room Temperature
Molecular Formula C9 H6 Cl2 N4 O
Certifications/Compliance ISO 17025
Synonym Lamotrigine Imp. A (EP), Lamotrigine USP Related Compound C, Lamotrigine USP RC C, 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one, 123W79, Ph Eur Lamotrigine Impurity A, USP Lamotrigine Related Compound C, 123W79 (Ph Eur Impurity A), (Ph Eur Lamotrigine Impurity A), Lamotrigine Related Compound C, Lamotrigine Impurity A
InChI Formula InChI=1S/C9H6Cl2N4O/c10-5-3-1-2-4(6(5)11)7-8(16)13-9(12)15-14-7/h1-3H,(H3,12,13,15,16)
SMILES NC1=NN=C(C(=O)N1)c2cccc(Cl)c2Cl
IUPAC Name 3-amino-6-(2,3-dichlorophenyl)-4H-1,2,4-triazin-5-one
Molecular Weight (g/mol) 257.08
Quantity 100 mg
Formula Weight 255.9919 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.