missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAMP3 Monoclonal Antibody (K24013_7C5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555819
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: TVSHTTGNTT QPSNQTTLPA TLSIALHKST TGQKPVQPTH APGTTAAAHN TTRTAAPAST VPGPTLAPQP SSVKTGIYQV LNGSRLCIKA EMGIQLIVQD KESVFSPRRY FNIDPNATQA SGNCGTRKSN
Dendritic cells (DCs) are the most potent antigen-presenting cells. Immature DCs efficiently capture antigens and differentiate into interdigitating dendritic cells (IDCs) in lymphoid tissues that induce primary T-cell responses (summary by de Saint-Vis et al., 1998 [PubMed 9768752]).
Specifications
| LAMP3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9UQV4 | |
| LAMP3 | |
| Recombinant human LAMP3 (immunogen range is 151-280). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K24013_7C5 | |
| Unconjugated | |
| LAMP3 | |
| 1200002D17Rik; CD208; DC LAMP; DCLAMP; DC-LAMP; DC-lysosome-associated membrane glycoprotein; dendritic cells-lysosome associated membrane glycoprotein; LAMP; Lamp3; LAMP-3; lysosomal associated membrane protein 3; lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; Protein TSC403; TSC403 | |
| Mouse | |
| Protein A | |
| RUO | |
| 27074 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction