missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LAMP3 Monoclonal Antibody (K24013_7C5), Invitrogen™
Mouse Monoclonal Antibody
438.00 € - 658.00 €
Specifications
| Antigen | LAMP3 |
|---|---|
| Clone | K24013_7C5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30205503
|
Invitrogen™
MA556199 |
200 μL |
658.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30202048
|
Invitrogen™
MA555819 |
50 μL |
438.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: TVSHTTGNTT QPSNQTTLPA TLSIALHKST TGQKPVQPTH APGTTAAAHN TTRTAAPAST VPGPTLAPQP SSVKTGIYQV LNGSRLCIKA EMGIQLIVQD KESVFSPRRY FNIDPNATQA SGNCGTRKSN
LAMP3 Monoclonal antibody specifically detects LAMP3 in Human samples. It is validated for Western BlotSpecifications
| LAMP3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 1200002D17Rik; CD208; DC LAMP; DCLAMP; DC-LAMP; DC-lysosome-associated membrane glycoprotein; dendritic cells-lysosome associated membrane glycoprotein; LAMP; Lamp3; LAMP-3; lysosomal associated membrane protein 3; lysosomal-associated membrane protein 3; lysosome-associated membrane glycoprotein 3; Protein TSC403; TSC403 | |
| Recombinant human LAMP3 (immunogen range is 151-280). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LAMP3 |
| K24013_7C5 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UQV4 | |
| 27074 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title