Learn More
Invitrogen™ LanthaScreen™ Elite Tb-anti-HIS Antibody
LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis.
1318.00 € - 17000.00 €
Specifications
Excitation/Emission 340/490, 520 nm
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10476768
View Documents
Invitrogen™
PV5863
25 μg
1318.00 €
25µg
View alternative products
Save Up to 
»
10326318
View Documents
Invitrogen™
PV5895
1 mg
17000.00 €
1mg
View alternative products
Save Up to 
»
Description
Description
LanthaScreen Elite Terbium-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using amine-reactive Tb chelates negatively impacts protein activity. The Elite Tb-anti-HIS-tag antibody is a specialized antibody with a broader HIS-tag binding performance than our traditional Tb-anti-HIS-tag antibody.
Specifications
Specifications
340/490, 520 nm