missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LanthaScreen™ Tb-PY20 Antibody Kit

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

Brand:  Thermo Scientific™ PV3552

 View more versions of this product

Product Code. 11553457

  • 920.00 € / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.

For use in combination with Fluorescein-poly-GT (Cat. No. PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tyrosine kinases
Tb (Terbium)/FITC/Lanthanide Chelates
25 μg
Kinase Antibodies and Immunoassays
1 Kit
150,000 Da
384-well Plate
Dry Ice
Signal Transduction
Tyrosine Kinases
ABL1, ABL2, AXL, BLK, BMX, BTK, CSF1R, CSK, DDR2, EGFR, EPHA1, EPHA2, EPHA3, EPHA4, EPHA5, EPHA7, EPHA8, EPHB1, EPHB2, EPHB3, EPHB4, ERBB2, ERBB4, FER, FGFR1, FGFR2, FGFR3, FGFR4, FGR, FLT1, FLT3, FLT4, FRK, FYN, HCK, IGF1R, INSR, KDR, KIT, LCK, LTK, LYNA, LYNB, MATK, MERTK, MET, MST1R, MUSK, NTRK1, NTRK2, NTRK3, PDGFRA, PDGFRB, PTK2, PTK2B, PTK6, RET, ROS1, SRC, SRCN1, SRMS, SYK, TEC, TEK, TNK2, TTK, TYRO3, YES1, ZAP70
0.45 mg/mL
150 kDa
RUO
Primary
Terbium-labeled anti-pY20 antibody in HEPES buffered saline. Store at -20°C.
Biochemical terbium phospho-specific activity
340/490, 520 nm
End Point
ABL1 G250E, CSF1R, CSK, EPHA5, KIT, PDGFRA T674I, PDGFRA V561D, PTK6, RET Y791F, SYK, TYRO3, ZAP70, FGFR1, FGFR2, BLK, EGFR, EPHB2, FGFR4, IGF1R, LYNA, MATK, MERTK, NTRK3, RET V804L, ROS1, TEK, TTK, FLT1, MUSK, SRCN1, TEC, ABL1 E255K, BMX, EPHA4, FYN, PTK2, PTK2B, RET, ABL1 T315I, EPHA3, EPHB4, FLT3, FLT4, INSR, LCK, YES1, EGFR L858R, ERBB4, KDR, MET, TNK2, ABL1, ABL2, AXL, FGFR3, FGFR3 K650E, FGR, HCK, KIT V654A, EPHA1, EPHA8, EPHB3, INSRR, LYNB, MST1R, NTRK1, PDGFRA D842V, SRMS, EGFR L861Q, EPHA2, FER, LTK, PDGFRA, PDGFRB, EGFR T790M, EPHA7, EPHB1, MET M1250T, FRK, ITK, FLT3 D835Y, SRC, ABL1 Y253F, BTK, DDR2, FES, KIT T670I, TXK, ERBB2, NTRK2
TR-FRET
LanthaScreen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.