Learn More
Thermo Scientific™ LanthaScreen™ Tb-PY20 Antibody Kit
A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
920.00 € - 14320.00 €
Specifications
Format 384-well Plate
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11553457
View Documents
Thermo Scientific™
PV3552
25 μg
920.00 €
25µg
View alternative products
Save Up to 
»
10042672
View Documents
Thermo Scientific™
PV3553
1 mg
14320.00 €
1mg
View alternative products
Save Up to 
»
Description
Description
A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.
For use in combination with Fluorescein-poly-GT (Cat. No. PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
384-well Plate