missing translation for 'onlineSavingsMsg'
Learn More

Latanoprost, Mikromol™

Product Code. 18032540
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18032540 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18032540 Supplier Mikromol Supplier No. MM3316.00
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Latanoprost
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Latanoprost
CAS 130209-82-4
Analyte or Component Names Latanoprost
Shipping Condition Room Temperature
Molecular Formula C26 H40 O5
Certifications/Compliance ISO 17025
Synonym 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]-, 1-methylethyl ester, (5Z)-, (15R)-Latanoprost, GAAP Ofteno, Latanoprost, PhXA 41, XA 41, Xalatan, 5-Heptenoic acid, 7-[3,5-dihydroxy-2-(3-hydroxy-5-phenylpentyl)cyclopentyl]-, 1-methylethyl ester, [1R-[1alpha(Z),2beta(R*),3alpha,5alpha]]-
InChI Formula InChI=1S/C26H40O5/c1-19(2)31-26(30)13-9-4-3-8-12-22-23(25(29)18-24(22)28)17-16-21(27)15-14-20-10-6-5-7-11-20/h3,5-8,10-11,19,21-25,27-29H,4,9,12-18H2,1-2H3/b8-3-/t21-,22+,23+,24-,25+/m0/s1
SMILES CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1CC[C@@H](O)CCc2ccccc2
IUPAC Name propan-2-yl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate
Molecular Weight (g/mol) 432.59
Quantity 50 mg
Formula Weight 432.2876 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.