missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latexin Antibody (8E1G6C1D4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06087-50ul
This item is not returnable.
View return policy
Description
Latexin Monoclonal antibody specifically detects Latexin in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Latexin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 8E1G6C1D4 | |
| Western Blot 1:500-1:1000, ELISA 1:5000-1:10000, Immunoprecipitation 1-2 uL/mg of lysate | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Latexin (Accession#: NP_064554.3; Glu 2-Glu 222). | |
| 50 μg | |
| Immunology, Innate Immunity | |
| 56925 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction