missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Latexin Antibody (8E1G6C1D4), Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 326.00 €
Specifications
| Antigen | Latexin |
|---|---|
| Clone | 8E1G6C1D4 |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:5000-1:10000, Immunoprecipitation 1-2 uL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654826
|
Novus Biologicals
NBP3-06087-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632618
|
Novus Biologicals
NBP3-06087-50ul |
50 μg |
220.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Latexin Monoclonal antibody specifically detects Latexin in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| Latexin | |
| Western Blot 1:500-1:1000, ELISA 1:5000-1:10000, Immunoprecipitation 1-2 uL/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Latexin (Accession#: NP_064554.3; Glu 2-Glu 222). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 8E1G6C1D4 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Immunology, Innate Immunity | |
| 0.2 um filtered solution in PBS | |
| 56925 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title