missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leucyl-cystinyl Aminopeptidase/LNPEP Antibody (E2) [Janelia Fluor« 646], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Leucyl-cystinyl Aminopeptidase/LNPEP Monoclonal antibody specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Leucyl-cystinyl Aminopeptidase/LNPEP |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | E2 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from human Leucyl-cystinyl Aminopeptidase/LNPEP (YFAATQFEPLAAR) |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?