missing translation for 'onlineSavingsMsg'
Learn More

Levocetirizine Dihydrochloride, LoGiCal

Product Code. 30462072
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462072 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462072 Supplier LoGiCal Supplier No. LGCFOR1035.00
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Levocetirizine Dihydrochloride
CAS 130018-87-0
Analyte or Component Names Levocetirizine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H25 Cl N2 O3 . 2 Cl H
Certifications/Compliance ISO 9001
Synonym Levocetirizine Dihydrochloride, Acetic acid, 2-[2-[4-[(R)-(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, hydrochloride (1:2), Acetic acid, [2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride, (R)-, Acetic acid, [2-[4-[(R)-(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, dihydrochloride (9CI), (R)-(-)-Cetirizine hydrochloride, (R)-Cetirizine dihydrochloride, Allear, Allercet, Levocetirizine dihydrochloride, Lezyncet, UCB 28556, Xusal, Xyzal, Xyzall
InChI Formula InChI=1S/C21H25ClN2O3.2ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H/t21-;;/m1../s1
Solution Type Neat
SMILES Cl.Cl.OC(=O)COCCN1CCN(CC1)[C@H](c2ccccc2)c3ccc(Cl)cc3
IUPAC Name 2-[2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 461.81
Quantity 10 mg
Formula Weight 460.1087 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.