missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Levofloxacin Methyl Ester, Mikromol™
Description
Levofloxacin Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Levofloxacin Methyl Ester |
| CAS | 862690-19-5 |
| Analyte or Component Names | Levofloxacin Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 F N3 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (3S)-9-Fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic acid methyl ester |
| InChI Formula | InChI=1S/C19H22FN3O4/c1-11-10-27-18-15-12(17(24)13(9-23(11)15)19(25)26-3)8-14(20)16(18)22-6-4-21(2)5-7-22/h8-9,11H,4-7,10H2,1-3H3/t11-/m0/s1 |
| SMILES | COC(=O)C1=CN2[C@@H](C)COc3c(N4CCN(C)CC4)c(F)cc(C1=O)c23 |
| Molecular Weight (g/mol) | 375.39 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?