missing translation for 'onlineSavingsMsg'
Learn More

LexA DNA Binding Region Antibody, Novus Biologicals™

Product Code. 18032904 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18032904 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18032904 Supplier Novus Biologicals Supplier No. NB6001137

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 2 publications

LexA DNA Binding Region Polyclonal specifically detects LexA DNA Binding Region in Bacteria samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LexA DNA Binding Region
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/ml, Immunoprecipitation 0.1 ug/ml
Formulation 0.1M Tris-Glycine (pH 7.4) and 0.15M NaCl with 0.05% Sodium Azide
Gene Alias ExrA; Lex A; LexA; LexA repressor; Spr; Tsl; UmuA
Host Species Rabbit
Immunogen His-tagged DNA binding domain of LexA protein corresponding to amino acid residues 1-202.
Molecular Weight of Antigen 23 kDa
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the LexA portion of bait fusion proteins used in the yeast two-hybrid system.
Target Species Bacteria
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.