missing translation for 'onlineSavingsMsg'
Learn More

LIF Antibody, Novus Biologicals™

Product Code. 18619297 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18619297 0.1 mg 0.1mg
18653009 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18619297 Supplier Novus Biologicals Supplier No. NBP311327

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIF Polyclonal antibody specifically detects LIF in Canine samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LIF
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Host Species Rabbit
Immunogen Recombinant canine Leukemia Inhibitory Factor
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines
Primary or Secondary Primary
Gene ID (Entrez) 3976
Target Species Canine
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.