missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Aconitate hydratase B Monoclonal specifically detects Aconitate hydratase B in E. coli samples. It is validated for Sandwich ELISA Capture.
Specifications
Specifications
| Antigen | Aconitate hydratase B |
| Applications | Sandwich ELISA Capture |
| Classification | Monoclonal |
| Clone | 09 |
| Conjugate | PerCP |
| Dilution | Sandwich ELISA Capture |
| Formulation | PBS |
| Gene Alias | acnB, hypothetical protein, yacI, yacJ |
| Host Species | Mouse |
| Immunogen | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Aconitate hydratase B (Uniprot#: P36683; Met1-Val865). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?