missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lipocalin-2/NGAL Antibody (32) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05182-0.025MG
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Lipocalin-2/NGAL | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:4000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3934 | |
| Rat | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, ELISA, Immunohistochemistry | |
| 32 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide | |
| LCN2 | |
| Recombinant rat NGAL precursor adsorbed onto aluminum hydroxide gel | |
| 0.025 mg | |
| Primary | |
| NGAL | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction