missing translation for 'onlineSavingsMsg'
Learn More

Lipocalin-2/NGAL Antibody (32) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

209.00 € - 591.00 €

Specifications

Antigen Lipocalin-2/NGAL
Clone 32
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, ELISA 1:4000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18700064
View Documents
Novus Biologicals
NBP1-05182
100 μg
591.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18256293
View Documents
Novus Biologicals
NBP1-05182-0.025MG
0.025 mg
209.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lipocalin-2/NGAL Monoclonal specifically detects Lipocalin-2/NGAL in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Lipocalin-2/NGAL
1.0 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Rat
3934
Recombinant rat NGAL precursor adsorbed onto aluminum hydroxide gel
Primary
Store at 4C in the dark.
32
Western Blot 1:100-1:2000, ELISA 1:4000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Monoclonal
Purified
RUO
24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGALlipocalin-2, Oncogene 24p3, p25, siderocalin
LCN2
IgG1 κ
Protein A or G purified
NGAL
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.