missing translation for 'onlineSavingsMsg'
Learn More

Lofexidine Hydrochloride, Mikromol™

Product Code. 30051706
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051706 Supplier Mikromol Supplier No. MM1898.00
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Lofexidine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lofexidine Hydrochloride
CAS 21498-08-8
Analyte or Component Names Lofexidine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C11 H12 Cl2 N2 O . Cl H
Certifications/Compliance ISO 17034
Synonym 1H-Imidazole, 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, hydrochloride (1:1), 1H-Imidazole, 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-, monohydrochloride (9CI), 2-Imidazoline, 2-[1-(2,6-dichlorophenoxy)ethyl]-, monohydrochloride (8CI), Baq 168, Britlofex, Lofetensin, Lofexidine hydrochloride, Loxacor, MDL 14042
InChI Formula InChI=1S/C11H12Cl2N2O.ClH/c1-7(11-14-5-6-15-11)16-10-8(12)3-2-4-9(10)13;/h2-4,7H,5-6H2,1H3,(H,14,15);1H
SMILES Cl.CC(Oc1c(Cl)cccc1Cl)C2=NCCN2
IUPAC Name 2-[1-(2,6-dichlorophenoxy)ethyl]-4,5-dihydro-1H-imidazole;hydrochloride
Molecular Weight (g/mol) 295.59
Quantity 100 mg
Formula Weight 294.0093 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.