missing translation for 'onlineSavingsMsg'
Learn More

Lomefloxacin Hydrochloride, Mikromol™

Product Code. 17904426
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17904426 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17904426 Supplier Mikromol Supplier No. MM0841.00
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Lomefloxacin Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lomefloxacin hydrochloride
CAS 98079-52-8
Analyte or Component Names Lomefloxacin hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H19 F2 N3 O3 . Cl H
Certifications/Compliance ISO 17025
Synonym Lomefloxacin Hydrochloride, 1-Ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxo-quinoline-3-carboxylic acid hydrochloride, 3-Quinolinecarboxylic acid, 1-ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, hydrochloride (1:1), 3-Quinolinecarboxylic acid, 1-ethyl-6,8-difluoro-1,4-dihydro-7-(3-methyl-1-piperazinyl)-4-oxo-, monohydrochloride (9CI), Alkaflox, Bareon, Chimono, Lomebact, Lomef 400, Lomflox, Lomitas, Lomoxin, Maxaquin, Mazaquin, NY 198, Orchacin, SC 47111, Uniquin
InChI Formula InChI=1S/C17H19F2N3O3.ClH/c1-3-21-8-11(17(24)25)16(23)10-6-12(18)15(13(19)14(10)21)22-5-4-20-9(2)7-22;/h6,8-9,20H,3-5,7H2,1-2H3,(H,24,25);1H
SMILES Cl.CCN1C=C(C(=O)O)C(=O)c2cc(F)c(N3CCNC(C)C3)c(F)c12
IUPAC Name 1-ethyl-6,8-difluoro-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 387.81
Quantity 100 mg
Formula Weight 387.1161 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.