missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loperamide.HCl, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Loperamide.HCl
Specifications
Specifications
| Chemical Name or Material | Loperamide Hydrochloride |
| CAS | 34552-83-5 |
| Analyte or Component Names | Loperamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H33 Cl N2 O2 . Cl H |
| Synonym | 4-(4-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinebutanamide monohydrochloride, 4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinobutyramide hydrochloride, Arret, Blox, Brek, Dissenten, Fortasec, Imodium, Imodium A-D, Imosec, Imossel, Lopemid, Lopemin, Loperamide hydrochloride, Loperazine, Loperyl, Lopodium, Maalox Antidiarrheal, NSC 696356, PJ 185, R 18553, Suprasec, Tebloc |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4 |
| IUPAC Name | 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction