missing translation for 'onlineSavingsMsg'
Learn More

Loratadine 0.1 mg/ml in Methanol, LoGiCal

Product Code. 30461609
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461609 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461609 Supplier LoGiCal Supplier No. LGCAMP0257.0002
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Loratadine 0.1 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Loratadine
CAS 79794-75-5
Analyte or Component Names Loratadine
Shipping Condition Room Temperature
Molecular Formula C22 H23 Cl N2 O2
Certifications/Compliance ISO 17034
Synonym 11 H-Benzo[5,6 ]cyclohepta[1,2-b]pyridine, 1-piperidinecarboxylic acid deriv., Alavert, Anhissen, Bonalerg, Civeran, Claratyne, Claritin, Claritine, Clarityn, Clarityne, Cronopen, Flonidan, Fristamin, Histaloran, Klaritin, Lertamine, Lisino, Loracert, Loradex, Loranox, Lorastine, Loratadine, Loratidine, Loratyne, Lorfast, Lowadina, Optimin, Polaratyne, Pylor, Restamine, Sch 29851, Sensibit, Sohotin, Tadine, Velodan, Zeos, Desloratadine Impurity C
InChI Formula InChI=1 S/C22H23ClN2O2/c1-2-27-22(26)25-12-9-15(10-13-25)20-19-8-7-18(23)14-17(19)6-5-16-4-3-11-24-21(16)20/h3-4,7-8,11,14 H,2,5-6,9-10,12-13H2,1H3
SMILES CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC1
IUPAC Name ethyl 4-(8-chloro-5,6-dihydrobenzo[1,2 ]cyclohepta[2,4-b]pyridin-11-ylidene)piperidine-1-carboxylate
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 382.88
Quantity 1 mL
Formula Weight 382.1448 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.