Learn More
LPAM-1 Rat, Biotin, Clone: DATK32, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB73-B-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15932418
  • 67.20 € / 25µg
Estimated Shipment: 26-01-2023
to see stock.
Specifications
LPAM-1
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Protein A/G Purified
RUO
Store at 2-4°C.
Flow Cytometry
DATK32
Biotin
Rat
25 μg
Primary
IgG2a κ