missing translation for 'onlineSavingsMsg'
Learn More

LRP-1 Antibody (1F6C6) - BSA Free, Novus Biologicals™

Product Code. p-200072580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.03mg
0.1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18108099 0.1 mg 0.1mg
18610018 0.025 mg 0.03mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18108099 Supplier Novus Biologicals Supplier No. NBP252507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LRP-1 Monoclonal specifically detects LRP-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Low Density LRP
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 1F6C6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias A2MRTGFBR5, Alpha-2-macroglobulin receptorMGC88725, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, FLJ16451, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, type V tgf-beta receptor
Gene Symbols LRP1
Host Species Mouse
Immunogen Purified recombinant fragment of human LRP1 (AA: 20-155) expressed in E. Coli.
Molecular Weight of Antigen 504 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4035
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.