missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21425-100ul
This item is not returnable.
View return policy
Description
LRP2 Polyclonal antibody specifically detects LRP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| LRP2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LRP2 | |
| 100 μg | |
| Lipid and Metabolism | |
| 4036 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction