missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
113.00 € - 363.00 €
Specifications
| Antigen | LRP2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18685404
|
Novus Biologicals
NBP3-21425-25ul |
25 μg |
113.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618584
|
Novus Biologicals
NBP3-21425-100ul |
100 μg |
363.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
LRP2 Polyclonal antibody specifically detects LRP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Spezifikation
| LRP2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| 0.2um filtered solution in PBS | |
| 4036 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human LRP2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts