missing translation for 'onlineSavingsMsg'
Learn More

LRRC8C Antibody - BSA Free, Novus Biologicals™

Product Code. 30556634 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30556634 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30556634 Supplier Novus Biologicals Supplier No. NBP346001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LRRC8C Polyclonal antibody specifically detects LRRC8C in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen LRRC8C
Applications ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:20-1:200, Immunocytochemistry/ Immunofluorescence 1:20-1:200
Formulation PBS, 50% Glycerol, pH (7.3)
Gene Alias AD158, DKFZp586J1119, factor for adipocyte differentiation 158, FAD158, leucine rich repeat containing 8 family, member C, leucine rich repeat containing 8C, leucine-rich repeat-containing protein 8C
Host Species Rabbit
Immunogen leucine rich repeat containing 8 family, member C. (Uniprot# Q8TDW0)
Purification Method Immunogen affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84230
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.