Learn More
LRRC8C Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-21601-20UL
Description
The LRRC8C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRRC8C. This antibody recognizes human, mouse, rat antigen. The LRRC8C antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| LRRC8C | |
| 1000 μg/mL | |
| AD158, DKFZp586J1119, FAD158, LRRC8C | |
| 20 μL | |
| 84230 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |