Learn More
LRRC8C Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | LRRC8C |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18087795
|
Proteintech
CL488-21601-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18026205
|
Proteintech
CL488-21601-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The LRRC8C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRRC8C. This antibody recognizes human, mouse, rat antigen. The LRRC8C antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| LRRC8C | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| AD158, DKFZp586J1119, FAD158, LRRC8C | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 84230 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.