missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM5 Mouse anti-Human, Clone: 25F8, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59399-100UL
This item is not returnable.
View return policy
Description
LSM5 Monoclonal specifically detects LSM5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| LSM5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1011, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| FLJ12710, LSM5 homolog, U6 small nuclear RNA associated (S. cerevisiae), U6 snRNA-associated Sm-like protein LSm5, YER146W | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 25F8 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| LSM5 | |
| Recombinant human LSM5 (1-91aa) purified from E. coli | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 23658 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction