missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSM5 Mouse anti-Human, Clone: 25F8, Novus Biologicals™
Mouse Monoclonal Antibody
294.00 € - 486.00 €
Specifications
| Antigen | LSM5 |
|---|---|
| Clone | 25F8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1011, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18258732
|
Novus Biologicals
NBP2-59399-50UL |
50 μL |
294.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18249263
|
Novus Biologicals
NBP2-59399-100UL |
100 μL |
486.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LSM5 Monoclonal specifically detects LSM5 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| LSM5 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Core ESC Like Genes, Stem Cell Markers | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 23658 | |
| Recombinant human LSM5 (1-91aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 25F8 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1011, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ12710, LSM5 homolog, U6 small nuclear RNA associated (S. cerevisiae), U6 snRNA-associated Sm-like protein LSm5, YER146W | |
| LSM5 | |
| IgG2a κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title