missing translation for 'onlineSavingsMsg'
Learn More

LSM8 Antibody [DyLight 488], Novus Biologicals™

Product Code. 30567454 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30567454 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30567454 Supplier Novus Biologicals Supplier No. NBP345992G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LSM8 Polyclonal antibody specifically detects LSM8 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LSM8
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias FLJ33242, LSM8 homolog, U6 small nuclear RNA associated (S. cerevisiae), MAK31-like protein, N(alpha)-acetyltransferase 38, NatC auxiliary subunit, N-alpha-acetyltransferase 38, NatC auxiliary subunit, U6 small nuclear RNA associated, YJR022W
Host Species Rabbit
Immunogen LSM8 homolog, U6 small nuclear RNA associated. (Uniprot# O95777)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51691
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.