missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LTB4R Recombinant Rabbit Monoclonal Antibody (24GB50)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557049

 View more versions of this product

Product Code. 30276580

  • 500.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

LTB4R Recombinant Monoclonal Antibody for Western Blot, ICC/IF

LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LTB4R
Recombinant Monoclonal
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O88855, Q9R0Q2
LTB4R, Ltb4r1
A synthesized peptide derived from human LTB4R (aa 322-352).
100 μL
Primary
Mouse, Rat
Antibody
IgG
Western Blot, Immunocytochemistry
24GB50
Unconjugated
Ltb4r1
BLT; BLT1; BLT-1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4 receptor; Ltb4r; LTB4-R 1; Ltb4r1; LTB4-R1; LTBR1; mBLTR; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7
Rabbit
Affinity chromatography
RUO
16995, 59264
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.