missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTB4R Recombinant Rabbit Monoclonal Antibody (24GB50)

Rabbit Recombinant Monoclonal Antibody
146.00 € - 500.00 €
Specifications
| Antigen | LTB4R |
|---|---|
| Clone | 24GB50 |
| Concentration | 0.25 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30276580
![]() |
Invitrogen™
MA557049 |
100 μL |
500.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275871
|
Invitrogen™
MA557050 |
20 μL |
146.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LTB4R Recombinant Monoclonal Antibody for Western Blot, ICC/IF
LTB4R (Leukotriene B4 Receptor) is a member of the GPCR family. It is a receptor for leukotriene B4, a potent chemoattractant involved in inflammation and immune response. It has been reported in blood, bone marrow, brain, heart, liver, and skeletal muscle. In addition, it has been shown to be highly expressed in leukocytes and lymphoid tissues such as spleen, thymus, and lymph node. ESTs have been isolated from B-cell/lung/testis, breast, colon, eye, kidney, skin, spleen, testis, and tonsil libraries.Specifications
| LTB4R | |
| 0.25 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88855, Q9R0Q2 | |
| 16995, 59264 | |
| A synthesized peptide derived from human LTB4R (aa 322-352). | |
| Antibody |
| 24GB50 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Mouse, Rat | |
| Ltb4r1 | |
| BLT; BLT1; BLT-1; BLTR; Chemoattractant receptor-like 1; chemokine receptor-like 1; CMKRL1; G protein-coupled receptor 16; GPR16; G-protein coupled receptor 16; leukotriene B4 receptor; leukotriene B4 receptor 1; LTB4 receptor; Ltb4r; LTB4-R 1; Ltb4r1; LTB4-R1; LTBR1; mBLTR; P2RY7; P2Y purinoceptor 7; P2Y7; purinergic receptor P2Y, G-protein coupled, 7 | |
| LTB4R, Ltb4r1 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title